Få mere at vide
Paraxanthine-1-methyl-d3, TRC
Artikelnummer. 30409681
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30409681 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30409681 Leverandør Toronto Research Chemicals Leverandørnr. TRCP1925021MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Paraxanthine-1-methyl-d3
CAS 188297-90-7
Renhedsgrad noter HPLC
Molekylær formel C7 2H3 H5 N4 O2
Mængde 1 mg
Synonym 1H-Purine-2,6-dione','3,7-dihydro-7-methyl-1-(methyl-d3)- (9CI), Paraxanthine-1-(methyl-d3)
InChI formel InChI=1S/C7H8N4O2/c1-10-3-8-5-4(10)6(12)11(2)7(13)9-5/h3H,1-2H3,(H,9,13)/i2D3
SMIL [2H]C([2H])([2H])N1C(=O)Nc2ncn(C)c2C1=O
IUPAC navn 7-methyl-1-(trideuteriomethyl)-3H-purine-2,6-dione
Molekylvægt (g/mol) 183.18
Formel vægt 183.0836
Procent renhed >95