missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Parathion-methyl, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Parathion-methyl |
| CAS | 298-00-0 |
| Molekylær formel | C8 H10 N O5 P S |
| Mængde | 500 mg |
| Synonym | Phosphorothioic acid','O,O-dimethyl O-(4-nitrophenyl) ester, Phosphorothioic acid','O,O-dimethyl O-(p-nitrophenyl) ester (6CI,8CI), 8056HC, Azofos, Azophos, Azophos (pesticide), BAY 11405, Bravik, Bravik 600CE, Dalf, Demethylfenitrothion, Dimethyl 4-nitrophenyl phosphorothioate, Dimethyl p-nitrophenyl phosphorothionate, Dimethyl p-nitrophenyl thiophosphate, Dimethyl parathion, Folidol 600, Folidol M, Folidol M 40, Folidol M 50, Folisuper 600BR, M-Parathion, ME 1605, ME 605, Me 605SP, Mentox 600CE, Meptox, Metacid, Metacid 50, Metacide, Metacide (insecticide), Metafos, Metafos (pesticide), Metaphos, Methion 48EC, Methyl 1605, Methyl 4E, Methyl Bladan, Methyl E 605, Methyl parathion, Methylthiophos, Metron, Metron (pesticide), Morphos, Nitrox, Nitrox 80, O,O-Dimethyl O-(4-nitrophenyl) phosphorothioate, O,O-Dimethyl O-(4-nitrophenyl) thiophosphate, O,O-Dimethyl O-(p-nitrophenyl) phosphorothioate, O,O-Dimethyl O-(p-nitrophenyl) thiophosphate, Oleovofotox, Parataf, Parathion M, Parathion methyl homolog, Parathion-methyl, Paratuf, Penncap M, Penncap MLS, Probel MP 2, Quinophos, Sinafid M 48, Thiophenit, Vofatox, Wofatox, Yphos |
| InChI formel | InChI=1S/C8H10NO5PS/c1-12-15(16,13-2)14-8-5-3-7(4-6-8)9(10)11/h3-6H,1-2H3 |
| SMIL | COP(=S)(OC)Oc1ccc(cc1)[N+](=O)[O-] |
| IUPAC navn | dimethoxy-(4-nitrophenoxy)-sulfanylidene-λ^{5}-phosphane |
| Molekylvægt (g/mol) | 263.21 |
| Formel vægt | 263.0017 |
Produkttitel
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.
Ser du en mulighed for forbedring?