Få mere at vide
Paramethasone Acetate, LoGiCal
Artikelnummer. 30457865
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30457865 10 mg
1 options
Artikelnummer. 30457865 Leverandør LoGiCal Leverandørnr. LGCFOR1667.00
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Paramethasone Acetate
CAS 1597-82-6
Analyt- eller komponentnavne Paramethasone Acetate
Forsendelsestilstand Room Temperature
Molekylær formel C24 H31 F O6
Certificeringer/compliance ISO 9001
Synonym Alondra, Cortidene, Dilar, Dillar, Flumethone, Haldrate, Haldrone, Metilar, Monocortin, Paramethasone 21-acetate, Paramethasone acetate, Paramezone, Stemex, Syntecort, Pregna-1,4-diene-3,20-dione, 21-(acetyloxy)-6-fluoro-11,17-dihydroxy-16-methyl-, (6alpha,11beta,16alpha)-, Pregna-1,4-diene-3,20-dione, 6alpha-fluoro-11beta,17,21-trihydroxy-16alpha-methyl-, 21-acetate (6CI,7CI,8CI), 16alpha-Methyl-6alpha-fluoroprednisolone 21-acetate, 21-Acetoxy-6alpha-fluoro-11beta,17-dihydroxy-16alpha-methylpregna-1,4-diene-3,20-dione, 21-Acetyl-6alpha-fluoro-16alpha-methylprednisolone, 6alpha-Fluoro-11beta,17,21-trihydroxy-16alpha-methylpregna-1,4-diene-3,20-dione 21-acetate, 6alpha-Fluoro-16alpha-methylprednisolone 21-acetate
InChI formel InChI=1S/C24H31FO6/c1-12-7-16-15-9-18(25)17-8-14(27)5-6-22(17,3)21(15)19(28)10-23(16,4)24(12,30)20(29)11-31-13(2)26/h5-6,8,12,15-16,18-19,21,28,30H,7,9-11H2,1-4H3/t12-,15+,16+,18+,19+,21-,22+,23+,24+/m1/s1
Løsningstype Neat
SMIL C[C@@H]1C[C@H]2[C@@H]3C[C@H](F)C4=CC(=O)C=C[C@]4(C)[C@H]3[C@@H](O)C[C@]2(C)[C@@]1(O)C(=O)COC(=O)C
IUPAC navn [2-[(6S,8S,9S,10R,11S,13S,14S,16R,17R)-6-fluoro-11,17-dihydroxy-10,13,16-trimethyl-3-oxo-7,8,9,11,12,14,15,16-octahydro-6H-cyclopenta[a]phenanthren-17-yl]-2-oxoethyl] acetate
Industry Type Clinical Testing
Molekylvægt (g/mol) 434.5
Mængde 10 mg
Formel vægt 434.2105 g/mol