Få mere at vide
Paracetamol EP Impurity J (Acetaminophen USP Related Compound J), Mikromol™
Beskrivelse
Paracetamol EP Impurity J (Acetaminophen USP Related Compound J)
Tekniske data
Tekniske data
| Kemisk navn eller materiale | 4-Chloroacetanilide |
| CAS | 539-03-7 |
| Analyt- eller komponentnavne | 4-Chloroacetanilide |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C8 H8 Cl N O |
| Certificeringer/compliance | ISO 17025 |
| Synonym | N-(4-Chlorophenyl)acetamide (4-Chloroacetanilide), N-(4-Chlorophenyl)acetamide, Paracetamol Imp. J (EP), 4-Chloroacetanilide, Acetaminophen USP Related Compound J, Acetaminophen USP RC J, SKF-8835, USP Acetaminophen Related Compound J, Ph Eur Paracetamol Impurity J, Acetaminophen Related Compound J, Paracetamol Impurity J |
| InChI formel | InChI=1S/C8H8ClNO/c1-6(11)10-8-4-2-7(9)3-5-8/h2-5H,1H3,(H,10,11) |
| SMIL | CC(=O)Nc1ccc(Cl)cc1 |
| IUPAC navn | N-(4-chlorophenyl)acetamide |
| Vis mere |