Få mere at vide
Paracetamol-D4 (Acetaminophen-D4) 1.0 mg/ml in Methanol, LoGiCal
Artikelnummer. 30462249
Regulatorisk: Dette produkt falder ind under Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. Hvis det er relevant, skal kunden fremskaffe en licens før levering.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30462249 Leverandør LoGiCal Leverandørnr. LGCAMP0042.8001
Regulatorisk: Dette produkt falder ind under Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. Hvis det er relevant, skal kunden fremskaffe en licens før levering.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Paracetamol-D4
CAS 64315-36-2
Opløsningsmiddel eller Matrix Methanol
Molekylær formel C8 2H4 H5 N O2
Mængde 1 mL
Synonym Paracetamol-D4, Acetaminophen-D4
InChI formel InChI=1S/C8H9NO2/c1-6(10)9-7-2-4-8(11)5-3-7/h2-5,11H,1H3,(H,9,10)/i2D,3D,4D,5D
SMIL [2H]c1c([2H])c(NC(=O)C)c([2H])c([2H])c1O
IUPAC navn N-(2,3,5,6-tetradeuterio-4-hydroxyphenyl)acetamide
Molekylvægt (g/mol) 155.19
Formel vægt 155.0884 g/mol