Få mere at vide
Para Red-d4, TRC
Artikelnummer. 30411680
Skift visning
Mængde:
50 mg
Artikelnummer. Mængde
30411680 50 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30411680 Leverandør Toronto Research Chemicals Leverandørnr. TRCP19197750MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Para Red-d4
CAS 1185235-75-9
Molekylær formel C16 2H4 H7 N3 O3
Mængde 50 mg
Synonym Para Red D4
InChI formel InChI=1S/C16H11N3O3/c20-15-10-5-11-3-1-2-4-14(11)16(15)18-17-12-6-8-13(9-7-12)19(21)22/h1-10,20H/b18-17+/i1D,3D,5D,10D
SMIL [2H]c1ccc2c(N=Nc3ccc(cc3)[N+](=O)[O-])c(O)c([2H])c([2H])c2c1[2H]
IUPAC navn 3,4,5,6-tetradeuterio-1-[(E)-(4-nitrophenyl)diazenyl]naphthalen-2-ol
Molekylvægt (g/mol) 297.3
Formel vægt 297.1051