missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Invitrogen™ PAR2 Monoclonal Antibody (SAM11)

Mouse Monoclonal Antibody
Brand: Invitrogen™ 352300
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
PAR2 Monoclonal Antibody for IHC, ELISA
Coagulation factor II (thrombin) receptor-like 1 (F2RL1) is a member of the large family of 7-transmembrane-region receptors that couple to guanosine-nucleotide-binding proteins. F2RL1 is also a member of the protease-activated receptor family. It is activated by trypsin, but not by thrombin. It is activated by proteolytic cleavage of its extracellular amino terminus. The new amino terminus functions as a tethered ligand and activates the receptor. The F2RL1 gene contains two exons and is widely expressed in human tissues. The predicted protein sequence is 83% identical to the mouse receptor sequence.
Tekniske data
| PAR2 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with 0.1% sodium azide; pH 7.4 | |
| P55085 | |
| F2rl1 | |
| A synthetic peptide corresponding to the N-terminal of the human PAR2 receptor. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
| ELISA, Immunohistochemistry | |
| SAM11 | |
| Unconjugated | |
| F2rl1 | |
| coagulation factor II (thrombin) receptor-like 1; coagulation factor II receptor-like 1; F2R like trypsin receptor 1; F2RL1; Gpcr11; Gpr11; G-protein coupled receptor 11; PAR2; PAR-2; protease-activated receptor 2; Protease-activated receptor-2; proteinase-activated receptor 2; Proteinase-activated receptor 2, alternate cleaved 1; Proteinase-activated receptor 2, alternate cleaved 2; proteinase-activated receptor-2; Proteinase-activated receptor-2 G protein-coupled receptor 11; Proteinase-activated receptor-2, G protein-coupled receptor 11; thrombin receptor-like 1 | |
| Mouse | |
| Protein A | |
| RUO | |
| 2150 | |
| -20°C | |
| Liquid |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion