Få mere at vide
MedChemExpress PAR-4 Agonist Peptide, amide
Artikelnummer. 16429538
Encompass_Preferred
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
16429538 10 mg
1 options
Artikelnummer. 16429538 Leverandør MedChemExpress Leverandørnr. HYP1309
MedChemExpress PAR-4 Agonist Peptide, amide (PAR-4-AP; AY-NH2) is a proteinase-activated receptor-4 (PAR-4) agonist, which has no effect on either PAR-1 or PAR-2 and whose effects are blocked by a PAR-4 antagonist.
Tekniske data
CAS 352017-71-1
Indhold og opbevaring Please store the product under the recommended conditions in the Certificate of Analysis.
Til brug med (applikation) COVID-19-immunoregulation
Molekylvægt (g/mol) 680.79 g/mol
Produkttype Peptides
Mængde 10 mg
Forsendelsestilstand Room temperature
Synonym PAR-4-AP∣ AY-NH2