Learn More
Paper Titan Yellow EU Pharmacopoeia, Fisher Chemical™
BIOPHARMA
Product Code. 12990554 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
6 Pk.
Unit Size:
50mL
Product Code. Quantity Unit Size
12990554 6 Pk. 50mL
1 options
Product Code. 12990554 Supplier Fisher Chemical Supplier No. RP 221 050
  • ONLINE EXCLUSIVE PRICE
    2345.00 DKK
    1759.33 DKK / Pakke med 50
    Spare 585.67 DKK
    24% Slukket
 Restricted Item
Forventet forsendelse: 14-01-2027
Pharmacopoeia reagents are intended for use in pharmacopoeia testing and monographs, produced under a GMP compliant ISO 9001 quality management system according to the relevant pharmacopoeia specification, using the pharmacopoeia reagent quality components and water meeting the relevant pharmacopoeia monograph requirement for Purified Water. Some pharmacopoeia standards are also accredited according to ISO 17025 and ISO Guide 43.
Specifications
Analyte or Component Names Paper Titan Yellow
Quantity 6 Pk.