Få mere at vide
Papaverine-d3 Hydrochloride, TRC
Artikelnummer. 30410442
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30410442 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30410442 Leverandør Toronto Research Chemicals Leverandørnr. TRCP19050210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Papaverine-d3
CAS 113718-66-4
Molekylær formel C20 2H3 H18 N O4 . Cl H
Mængde 10 mg
InChI formel InChI=1S/C20H21NO4.ClH/c1-22-17-6-5-13(10-18(17)23-2)9-16-15-12-20(25-4)19(24-3)11-14(15)7-8-21-16;/h5-8,10-12H,9H2,1-4H3;1H/i4D3;
SMIL Cl.[2H]C([2H])([2H])Oc1cc2c(Cc3ccc(OC)c(OC)c3)nccc2cc1OC
IUPAC navn 1-[(3,4-dimethoxyphenyl)methyl]-6-methoxy-7-(trideuteriomethoxy)isoquinoline;hydrochloride
Molekylvægt (g/mol) 378.86
Formel vægt 378.1426