Få mere at vide
Pantothenic Acid (Technical Grade, Contains Lactone), TRC
Artikelnummer. 30132771
Skift visning
Mængde:
250 mg
2.5 g
Pakningsstørrelse:
2.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30132771 2.5 g 2.5g
30011170 250 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30132771 Leverandør Toronto Research Chemicals Leverandørnr. TRCP1903052.5G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 79-83-4
Molekylær formel C9 H17 N O5
Molekylvægt (g/mol) 219.24
Synonym β-Alanine, N-[(2R)-2,4-dihydroxy-3,3-dimethyl-1-oxobutyl]-, Pantothenic acid, D- (8CI), β-Alanine, N-(2,4-dihydroxy-3,3-dimethyl-1-oxobutyl)-, (R)-, (+)-Pantothenic acid, (D)-(+)-Pantothenic acid, Chick antidermatitis factor, D(+)-N-(2,4-Dihydroxy-3,3-dimethylbutyryl)-β-alanine, D-Pantothenic acid, N-[(2R)-2,4-Dihydroxy-3,3-dimethyl-1-oxobutyl]-β-alanine, Pantothenic acid, Vitamin B3, Vitamin B5
IUPAC navn 3-[[(2R)-2,4-dihydroxy-3,3-dimethylbutanoyl]amino]propanoic acid
SMIL CC(C)(CO)[C@@H](O)C(=O)NCCC(=O)O
Tekniske data
Kemisk navn eller materiale D-Pantothenic acid
Mængde 2.5 g
InChI formel InChI=1S/C9H17NO5/c1-9(2,5-11)7(14)8(15)10-4-3-6(12)13/h7,11,14H,3-5H2,1-2H3,(H,10,15)(H,12,13)/t7-/m0/s1
Formel vægt 219.1107