Få mere at vide
Pantothenic Acid-13C3,15N Hemicalcium Salt, TRC
Artikelnummer. 17940095
Skift visning
Mængde:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17940095 10 mg
17938664 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 17940095 Leverandør Toronto Research Chemicals Leverandørnr. TRCP19030210MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 356786-94-2
Molekylær formel 2 13C3 C6 H16 15N O5 . Ca
Molekylvægt (g/mol) 484.47
Synonym β-Alanine-1,2,3-13C3-15N, N-[(2R)-2,4-dihydroxy-3,3-dimethyl-1-oxobutyl]-, calcium salt (2:1), β-Alanine-13C3-15N, N-[(2R)-2,4-dihydroxy-3,3-dimethyl-1-oxobutyl]-, calcium salt (2:1) (9CI), Vitamin B5 (β-alanine-13C3,15N) calcium salt, Vitamin B5-[13C6,15N2], Calcium Pantothenate-[13C6,15N2], Pantothenic acid-13C3,15N hemicalcium salt, D-Pantothenic acid-13C3,15N hemicalcium salt (β-alanine-13C3,15N)
IUPAC navn calcium;3-[[(2R)-2,4-dihydroxy-3,3-dimethylbutanoyl]amino](1,2,3-^{13}C_{3})propanoate
SMIL [Ca+2].CC(C)(CO)[C@@H](O)C(=O)[15NH][13CH2][13CH2][13C](=O)[O-].CC(C)(CO)[C@@H](O)C(=O)[15NH][13CH2][13CH2][13C](=O)[O-]
Tekniske data
Kemisk navn eller materiale D-Pantothenic acid-13C3,15N hemicalcium salt
Renhedsgrad noter HPLC
Mængde 10 mg
InChI formel InChI=1S/2C9H17NO5.Ca/c2*1-9(2,5-11)7(14)8(15)10-4-3-6(12)13;/h2*7,11,14H,3-5H2,1-2H3,(H,10,15)(H,12,13);/q;;+2/p-2/t2*7-;/m00./s1/i2*3+1,4+1,6+1,10+1;
Formel vægt 484.1825
Procent renhed >95%