Få mere at vide
Pantoprazole Sulfide-D7 (Major), TRC
Artikelnummer. 30425874
Skift visning
Mængde:
50 mg
Artikelnummer. Mængde
30425874 50 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30425874 Leverandør Toronto Research Chemicals Leverandørnr. TRCP18302250MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Pantoprazole Sulfide-D7 (Major)
CAS 1189977-42-1
Molekylær formel C16 D8 H7 F2 N3 O3 S
Mængde 50 mg
Synonym Pantoprazole Thioether-D6, 5-(Difluoromethoxy)-2-[[(3,4-dimethoxy-2-pyridinyl)methyl]thio]-1H-benzimidazole-D6
InChI formel InChI=1S/C16H15F2N3O3S/c1-22-13-5-6-19-12(14(13)23-2)8-25-16-20-10-4-3-9(24-15(17)18)7-11(10)21-16/h3-7,15H,8H2,1-2H3,(H,20,21)/i1D3,2D,5D,6D,8D2
SMIL [2H]COc1c(OC([2H])([2H])[2H])c([2H])c([2H])nc1C([2H])([2H])Sc2nc3cc(OC(F)F)ccc3[nH]2
IUPAC navn 2-[dideuterio-[5,6-dideuterio-3-(deuteriomethoxy)-4-(trideuteriomethoxy)pyridin-2-yl]methyl]sulfanyl-5-(difluoromethoxy)-1H-benzimidazole
Molekylvægt (g/mol) 375.42
Formel vægt 375.13