Få mere at vide
Panobinostat(LBH-589), TRC
Artikelnummer. 30411139
Skift visning
Mængde:
50 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30411139 50 mg
30414768 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30411139 Leverandør Toronto Research Chemicals Leverandørnr. TRCP18050050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Panobinostat
CAS 404950-80-7
Molekylær formel C21 H23 N3 O2
Mængde 50 mg
Synonym 2-Propenamide','N-hydroxy-3-[4-[[[2-(2-methyl-1H-indol-3-yl)ethyl]amino]methyl]phenyl]-','(2E)-, (2E)-N-Hydroxy-3-[4-[[[2-(2-methyl-1H-indol-3-yl)ethyl]amino]methyl]phenyl]-2-propenamide, Farydak, LBH 589, Panobinostat
InChI formel InChI=1S/C21H23N3O2/c1-15-18(19-4-2-3-5-20(19)23-15)12-13-22-14-17-8-6-16(7-9-17)10-11-21(25)24-26/h2-11,22-23,26H,12-14H2,1H3,(H,24,25)/b11-10+
SMIL Cc1[nH]c2ccccc2c1CCNCc3ccc(\C=C\C(=O)NO)cc3
IUPAC navn (E)-N-hydroxy-3-[4-[[2-(2-methyl-1H-indol-3-yl)ethylamino]methyl]phenyl]prop-2-enamide
Molekylvægt (g/mol) 349.43
Formel vægt 349.179