Få mere at vide
Panidazole, LoGiCal
Artikelnummer. 30460867
Skift visning
Mængde:
5 mg
Artikelnummer. Mængde
30460867 5 mg
1 options
Artikelnummer. 30460867 Leverandør LoGiCal Leverandørnr. LGCFOR3168.00
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Panidazole
CAS 13752-33-5
Analyt- eller komponentnavne Panidazole
Forsendelsestilstand Room Temperature
Molekylær formel C11 H12 N4 O2
Certificeringer/compliance ISO 9001
Synonym Pyridine, 4-[2-(2-methyl-5-nitro-1H-imidazol-1-yl)ethyl]-, Pyridine, 4-[2-(2-methyl-5-nitroimidazol-1-yl)ethyl]- (8CI), 2-Methyl-5-nitro-1-[2-(4-pyridyl)ethyl]imidazole, Panidazole
InChI formel InChI=1S/C11H12N4O2/c1-9-13-8-11(15(16)17)14(9)7-4-10-2-5-12-6-3-10/h2-3,5-6,8H,4,7H2,1H3
Løsningstype Neat
SMIL Cc1ncc([N+](=O)[O-])n1CCc2ccncc2
IUPAC navn 4-[2-(2-methyl-5-nitroimidazol-1-yl)ethyl]pyridine
Industry Type Clinical Testing
Molekylvægt (g/mol) 232.24
Mængde 5 mg
Formel vægt 232.096 g/mol