missing translation for 'onlineSavingsMsg'
Learn More
Pamapimod, MedChemExpress Product Code.: 16433948

Pamapimod, MedChemExpress

Product Code. p-200101690
10 mg, 10mg
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
This item is not returnable. View return policy

Product Code. 16433948

Brand: Medchem Express HY10405

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Pamapimod (Ro4402257) is a potent, selective and orally active p38 MAPK inhibitor with IC50s of 14 nM and 480 nM and Kis of 1.3 nM and 120 nM for p38α and p38β, respectively. Pamapimod has no activity against p38δ or p38γ isoforms. Pamapimod has the potential for rheumatoid arthritis and other autoimmune diseases treatment.

Pamapimod (Ro4402257) is a potent, selective and orally active p38 MAPK inhibitor with IC50s of 14 nM and 480 nM and Kis of 1.3 nM and 120 nM for p38α and p38β, respectively. Pamapimod has no activity against p38δ or p38γ isoforms. Pamapimod has the potential for rheumatoid arthritis and other autoimmune diseases treatment.

Specifications

Chemical Name or Material Pamapimod
CAS 449811-01-2
Color Off-White
Purity Grade Notes Research
Molecular Formula C19H20F2N4O4
Quantity 10 mg
Synonym Ro4402257 R1503
Solubility Information DMSO : ≥ 34 mg/mL (83.67 mM)
SMILES O=C1C(OC2=CC=C(F)C=C2F)=CC3=CN=C(NC(CCO)CCO)N=C3N1C
Molecular Weight (g/mol) 406.38
Formula Weight 406.38
Percent Purity 99.92%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title
Pamapimod, MedChemExpress

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.