Få mere at vide
Palustric Acid, TRC
Artikelnummer. 30008813
Skift visning
Mængde:
2.5 mg
25 mg
250 mg
Pakningsstørrelse:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30008813 250 mg -
30008968 25 mg -
17902525 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30008813 Leverandør Toronto Research Chemicals Leverandørnr. TRCP165950250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1945-53-5
Molekylær formel C20 H30 O2
Molekylvægt (g/mol) 302.45
Synonym 1-Phenanthrenecarboxylic acid, 1,2,3,4,4a,5,6,9,10,10a-decahydro-1,4a-dimethyl-7-(1-methylethyl)-, [1R-(1α,4aβ,10aα)]-, Palustric acid (6CI), Podocarpa-8,13-dien-15-oic acid, 13-isopropyl- (8CI), (1R,4aS,10aR)-1,2,3,4,4a,5,6,9,10,10a-Decahydro-1,4a-dimethyl-7-(1-methylethyl)-1-phenanthrenecarboxylic acid, 8,13-Abietadien-18-oic acid, NSC 148945
IUPAC navn (1R,4aS,10aR)-1,4a-dimethyl-7-propan-2-yl-2,3,4,5,6,9,10,10a-octahydrophenanthrene-1-carboxylic acid
SMIL CC(C)C1=CC2=C(CC1)[C@@]3(C)CCC[C@](C)([C@@H]3CC2)C(=O)O
Tekniske data
Kemisk navn eller materiale Palustric Acid
Mængde 250 mg
InChI formel InChI=1S/C20H30O2/c1-13(2)14-6-8-16-15(12-14)7-9-17-19(16,3)10-5-11-20(17,4)18(21)22/h12-13,17H,5-11H2,1-4H3,(H,21,22)/t17-,19-,20-/m1/s1
Formel vægt 302.2246