missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Palonosetron Hydrochlorid, Mikromol™
Description
Palonosetron Hydrochlorid
Specifications
Specifications
| Kemisk navn eller materiale | Palonosetron Hydrochloride |
| CAS | 135729-62-3 |
| Analyt- eller komponentnavne | Palonosetron Hydrochloride |
| Forsendelsestilstand | Stuetemperatur |
| Molekylær formel | C19 H24 N2 O . Cl H |
| Certificeringer/compliance | ISO 17034 |
| Synonym | 1H-Benz[de]isoquinolin-1-one, 2-[(3S)-1-azabicyclo[2.2.2]oct-3-yl]-2,3,3a,4,5,6-hexahydro-, hydrochloride (1:1), (3aS)-, 1H-Benz[de]isoquinolin-1-one, 2-(1-azabicyclo[2.2.2]oct-3-yl)-2,3,3a,4,5,6-hexahydro-, monohydrochloride, [S-(R*,R*)]-, 1H-Benz[de]isoquinolin-1-one, 2-(3S)-1-azabicyclo[2.2.2]oct-3-yl-2,3,3a,4,5,6-hexahydro-, monohydrochloride, (3aS)- (9CI), Aloxi, Onicit, Palonosetron hydrochloride, RS 25259-197 |
| InChI formel | InChI=1S/C19H24N2O.ClH/c22-19-16-6-2-4-14-3-1-5-15(18(14)16)11-21(19)17-12-20-9-7-13(17)8-10-20;/h2,4,6,13,15,17H,1,3,5,7-12H2;1H/t15-,17-;/m1./s1 |
| SMIL | Cl.O=C1N(C[C@H]2CCCc3cccc1c23)[C@@H]4CN5CCC4CC5 |
| IUPAC navn | (3aS)-2-[(3S)-1-azabicyclo[2.2.2]oktan-3-yl]-3a,4,5,6-tetrahydro-3H-benzo[de]isoquinolin-1-one; hydrochlorid |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?