Få mere at vide
Palmitoylethanolamide-d4, TRC
Artikelnummer. 30427765
Skift visning
Mængde:
5 mg
Artikelnummer. Mængde
30427765 5 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30427765 Leverandør Toronto Research Chemicals Leverandørnr. TRCP1550825MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Palmitoylethanolamide-d4
CAS 946524-34-1
Molekylær formel C18 D4 H33 N O2
Mængde 5 mg
Synonym N-(2-Hydroxyethyl-1,1,2,2-d4)hexadecanamide, (Hydroxyethyl-d4)palmitamide, 2-(Palmitoylamino)ethanol-d4, 2-Palmitamidoethanol-d4, AM 3112-d4, Impulsin-d4, Loramine P 256-d4, Mackpeart DR 14V-d4, N-(2-Hydroxyethyl-d4)hexadecanamide, N-(2-Hydroxyethyl-d4)palmitamide, N-Hexadecanoylethanol-d4-amine, N-Palmitoylethanol-d4-amine, NSC 23320-d4, Palmidrol-d4, Palmitic Acid Monoethanol-d4-amide, Palmitic Monoethanol-d4-amide
InChI formel InChI=1S/C18H37NO2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-18(21)19-16-17-20/h20H,2-17H2,1H3,(H,19,21)/i16D2,17D2
SMIL [2H]C([2H])(O)C([2H])([2H])NC(=O)CCCCCCCCCCCCCCC
IUPAC navn N-(1,1,2,2-tetradeuterio-2-hydroxyethyl)hexadecanamide
Molekylvægt (g/mol) 303.517
Formel vægt 303.308