Få mere at vide
Palmitoylcholine Chloride, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Palmitoylcholine Chloride |
| CAS | 2932-74-3 |
| Molekylær formel | C21 H44 N O2 . Cl |
| Mængde | 2.5 g |
| Synonym | N-[β-(Hexadecanoyloxy)ethyl]trimethylammonium Chloride, Palmitic Acid','Ester with Choline Chloride, Choline Palmitate Chloride, N-[β-(Hexadecanoyloxy)ethyl]trimethylammonium Chloride, PM 2MC, Palmitoylcholine Chloride |
| InChI formel | InChI=1S/C21H44NO2.Cl/c1-5-6-7-8-9-10-11-12-13-14-15-16-17-18-21(23)24-20-19-22(2,3)4;/h5-20H2,1-4H3;/q+1; |
| SMIL | [Cl].CCCCCCCCCCCCCCCC(=O)OCC[N+](C)(C)C |
| IUPAC navn | chlorine;2-hexadecanoyloxyethyl(trimethyl)azanium |
| Molekylvægt (g/mol) | 378.033 |
| Formel vægt | 377.306 |