Få mere at vide
Palmitoyl-L-carnitine-d3 Hydrochloride, TRC
Artikelnummer. 30420465
Skift visning
Mængde:
2.5 mg
Pakningsstørrelse:
2.5mg
Artikelnummer. Mængde unitSize
30420465 2.5 mg 2.5mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30420465 Leverandør Toronto Research Chemicals Leverandørnr. TRCP1550522.5MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Palmitoyl-L-carnitine-d3 Hydrochloride
CAS 1334532-26-1
Molekylær formel C23 D3 H43 N O4 . Cl
Mængde 2.5 mg
Synonym (2R)-3-Carboxy-N,N,N-trimethyl-2-[(1-oxohexadecyl)oxy]-1-propanaminium-d3 Chloride, L-(3-Carboxy-2-hydroxypropyl)trimethyl-ammonium-d3 Chloride Palmitate, L-Palmitic acid ester with (3-carboxy-2-hydroxypropyl)trimethylammonium-d3 chloride, (-)-(3-Carboxy-2-hydroxypropyl)trimethyl-ammonium-d3 Chloride Palmitate, Hexadecanoyl-L-carnitine-d3 hydrochloride, L-Palmitoylcarnitine-d3 chloride, Hexadecanoyl-L-carnitine-d3 HCl (N-methyl-d3)
InChI formel InChI=1S/C23H45NO4.ClH/c1-5-6-7-8-9-10-11-12-13-14-15-16-17-18-23(27)28-21(19-22(25)26)20-24(2,3)4;/h21H,5-20H2,1-4H3;1H/t21-;/m1./s1/i2D3;
SMIL [Cl-].[2H]C([2H])([2H])[N+](C)(C)C[C@@H](CC(=O)O)OC(=O)CCCCCCCCCCCCCCC
IUPAC navn [(2R)-3-carboxy-2-hexadecanoyloxypropyl]-dimethyl-(trideuteriomethyl)azanium;chloride
Molekylvægt (g/mol) 439.09
Formel vægt 438.3304