Få mere at vide
Palmitic Acid Methylamide, TRC
Artikelnummer. 30426759
Skift visning
Mængde:
250 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30426759 500 mg
30410742 1 g
30422393 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30426759 Leverandør Toronto Research Chemicals Leverandørnr. TRCP144508500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 7388-58-1
Molekylær formel C17H35NO
Molekylvægt (g/mol) 269.47
Synonym N-Methyl-hexadecanamide, N-Methylhexadecanamide, N-Methylpalmitamide
IUPAC navn N-methylhexadecanamide
SMIL CCCCCCCCCCCCCCCC(=O)NC
Tekniske data
Kemisk navn eller materiale Palmitic Acid Methylamide
Mængde 500 mg
InChI formel InChI=1S/C17H35NO/c1-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17(19)18-2/h3-16H2,1-2H3,(H,18,19)
Formel vægt 269.2719