Få mere at vide
Palmitic Acid-d4, TRC
Artikelnummer. 30421554
Skift visning
Mængde:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30421554 5 mg
30410646 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30421554 Leverandør Toronto Research Chemicals Leverandørnr. TRCP1445025MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 75736-47-9
Molekylær formel C16 D4 H28 O2
Molekylvægt (g/mol) 260.449
Synonym Hexadecanoic-5,5,6,6-d4 acid (9CI), Hexadecanoic-5,5,6,6-D4 acid, [5,5,6,6-2H4]Hexadecanoic acid, Hexadecanoic acid-5,5,6,6-d4, Palmitic-5,5,6,6-d4 acid, Palmitic acid-5,5,6,6-d4
IUPAC navn 5,5,6,6-tetradeuteriohexadecanoic acid
SMIL [2H]C([2H])(CCCCCCCCCC)C([2H])([2H])CCCC(=O)O
Tekniske data
Kemisk navn eller materiale Palmitic Acid-d4
Mængde 5 mg
InChI formel InChI=1S/C16H32O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16(17)18/h2-15H2,1H3,(H,17,18)/i11D2,12D2
Formel vægt 260.265