Få mere at vide
Palmitic Acid-d3, TRC
Artikelnummer. 30426063
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30426063 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30426063 Leverandør Toronto Research Chemicals Leverandørnr. TRCP14450310MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Palmitic Acid-d3
CAS 75736-53-7
Molekylær formel C16 D3 H29 O2
Mængde 10 mg
Synonym Hexadecanoic-16,16,16-d3 acid (9CI','ACI), [16,16,16-2H3]Hexadecanoic acid, Hexadecanoic-16,16,16-d3 acid, Hexadecanoic acid-16,16,16-d3, Palmitic-16,16,16-d3 acid, Palmitic-ω-d3 acid
InChI formel InChI=1S/C16H32O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16(17)18/h2-15H2,1H3,(H,17,18)/i1D3
SMIL [2H]C([2H])([2H])CCCCCCCCCCCCCCC(=O)O
IUPAC navn 16,16,16-trideuteriohexadecanoic acid
Molekylvægt (g/mol) 259.44
Formel vægt 259.2591