Få mere at vide
Palmitic Acid-13C16, TRC
Artikelnummer. 30414929
Skift visning
Mængde:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30414929 10 mg
30411974 5 mg
30409304 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30414929 Leverandør Toronto Research Chemicals Leverandørnr. TRCP14450510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 56599-85-0
Molekylær formel 13C16 H32 O2
Molekylvægt (g/mol) 272.31
Synonym Hexadecanoic-13C16 Acid
IUPAC navn (1,2,3,4,5,6,7,8,9,10,11,12,13,14,15,16-^{13}C_{16})hexadecanoic acid
SMIL O[13C]([13CH2][13CH2][13CH2][13CH2][13CH2][13CH2][13CH2][13CH2][13CH2][13CH2][13CH2][13CH2][13CH2][13CH2][13CH3])=O
Tekniske data
Kemisk navn eller materiale Palmitic Acid-13C16
Mængde 10 mg
InChI formel InChI=1S/C16H32O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16(17)18/h2-15H2,1H3,(H,17,18)/i1+1,2+1,3+1,4+1,5+1,6+1,7+1,8+1,9+1,10+1,11+1,12+1,13+1,14+1,15+1,16+1
Formel vægt 272.2939