missing translation for 'onlineSavingsMsg'
Learn More
Palladium, plasma standard solution, Specpure™ Pd 1000μg/mL Product Code.: 11351389

Palladium, plasma standard solution, Specpure™ Pd 1000μg/mL

Product Code. p-7082599
50 mL, 50mL
Click to view available options
Quantity:
50 mL
100 mL
500 mL
Unit Size:
100mL
500mL
50mL
This item is not returnable. View return policy

Product Code. 11351389

Brand: Thermo Scientific Alfa Aesar 013833.AD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Pd in 20% HCl

Palladium, plasma standard solution finds application as a single-element standard solution for plasma emission spectrometry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

Molecular Formula Pd in 20% HCl
MDL Number MFCD03613603

Specifications

Physical Form Liquid
Concentration Pd 1000 ÎĽg/mL
UN Number UN1789
Solubility Miscible with water
Quantity 50 mL
Chemical Name or Material Palladium, plasma standard solution, Specpure™

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Product Code
Lot Number1523444 Certificate TypeCertificate of Analysis Date13/03/2025 Product Code013833.AP,013833.AD,013833.AE
Lot Number1528129 Certificate TypeCertificate of Analysis Date13/03/2025 Product Code013833.AP,013833.AD,013833.AE
Lot Number1528130 Certificate TypeCertificate of Analysis Date13/03/2025 Product Code013833.AP,013833.AD,013833.AE
Lot Number1528122 Certificate TypeCertificate of Analysis Date13/03/2025 Product Code013833.AP,013833.AD,013833.AE
Lot Number1517374 Certificate TypeCertificate of Analysis Date15/01/2025 Product Code013833.AP,013833.AD,013833.AE

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Palladium, plasma standard solution, Specpure™ Pd 1000μg/mL >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.