missing translation for 'onlineSavingsMsg'
Få mere at vide

Palladium(II) nitrate dihydrate (40% Pd), Sigma-Aldrich™

Artikelnummer. 32238868
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
32238868 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 32238868 Supplier Sigma Aldrich Supplier No. 8145738500
 Begrænset vare
Forventet forsendelse: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

For synthesis

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale Palladium(II) nitrate dihydrate (40% Pd)
CAS 32916-07-7
Mængde 500 mg
Molekylær formel Pd(NO3)2 · 2 H2O
Synonym Palladium(II) nitrate dihydrate (40% Pd), Palladium(II) nitrate dihydrate (40% Pd)
Molekylvægt (g/mol) 266.46
Grad Not Available
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.