Få mere at vide
Palladium(II) Acetylacetonate, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Palladium(II) Acetylacetonate |
| CAS | 14024-61-4 |
| Molekylær formel | 2 C5 H7 O2 . Pd |
| Mængde | 2.5 g |
| Synonym | (SP-4-1)-Bis(2,4-pentanedionato-κO2,κO4)palladium, Bis(2,4-pentanedionato)palladium, Bis(2,4-pentanedionato-κO,κO')palladium, Bis(acetylacetonato)palladium, Bis(acetylacetonato)palladium(II), NSC 187660, Palladium Acetylacetonate, Palladium bis(2,4-pentanedionate), Palladium Bis(acetoacetonate), Palladium Bis(acetylacetonate), Palladium(II) Acetylacetonate, Pd (II) Diacetylacetonate, Bis(2,4-pentanedionato-κO,κO')-','(SP-4-1)-Palladium','Bis(2,4-pentanedionato)-palladium |
| InChI formel | InChI=1S/2C5H7O2.Pd/c2*1-4(6)3-5(2)7;/h2*3H,1-2H3;/q2*-1;+2 |
| SMIL | [Pd+2].CC(=O)[CH-]C(=O)C.CC(=O)[CH-]C(=O)C |
| IUPAC navn | palladium(2+);pentane-2,4-dione |
| Molekylvægt (g/mol) | 304.636 |
| Formel vægt | 303.993 |