missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Palladium, 5% on carbon paste, sulfided, Type A103023-5

Brand:  Alfa Aesar™ H37400.06

Additional Details : CAS Number : 7440-05-3 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15407767

  • 106.42 DKK / 5g


Estimated Shipment date
from Supplier 01-08-2018
Add to basket

Specifications

Chemical Identifiers

Palladium
7440-05-3
5% on carbon paste, sulfided, Type A103023-5
MFCD03457879

Specifications

Palladium
7440-05-3
MFCD03457879
Black
5% on carbon paste, sulfided, Type A103023-5
Powder
Odorless
5g
Videos
SDS