missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Palladium, 5% on activated carbon powder, Type 487

Brand:  Alfa Aesar™ H37587.22

Additional Details : CAS Number : 7440-05-3 Weight : 0.10000kg Transport : UN number : 1362 Chem class : 4.2 Pack group : III

Quantity 100g

 View more versions of this product

Product Code. 15477797

  • 5806.83 DKK / 100g


Estimated Shipment
06-10-2020
Add to basket

Specifications

Chemical Identifiers

7440-05-3
106.42
KDLHZDBZIXYQEI-UHFFFAOYSA-N
23938
palladium
Pd
MFCD03457879
black, on carbon, paladio, palladium/carbon, element, charcoal, palladium, element, palladex 600, on charcoal, palladium-activated carbon
CHEBI:33363
[Pd]

Specifications

Palladium
7440-05-3
MFCD03457879
black, on carbon, paladio, palladium/carbon, element, charcoal, palladium, element, palladex 600, on charcoal, palladium-activated carbon
[Pd]
106.42
CHEBI:33363
Odorless
100g
5% on activated carbon powder, Type 487
Pd
UN1362
KDLHZDBZIXYQEI-UHFFFAOYSA-N
palladium
23938
Powder
Gray
Videos
SDS Documents