Learn More
Paliperidone-d4, TRC
Product Code. 30420315
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
Product Code. Quantity Unit Size
30420315 1 mg 1mg
1 options
Regulatory: This product fall under regulation “Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. If applicable, customers must provide a license before delivery.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30420315 Supplier Toronto Research Chemicals Supplier No. TRCP1410021MG
 Restricted Item
Forventet forsendelse: 10-12-2026
Regulatory: This product fall under regulation “Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. If applicable, customers must provide a license before delivery.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material rac 9-Hydroxy Risperidone-d4
CAS 1020719-55-4
Purity Grade Notes HPLC
Molecular Formula C23 2H4 H23 F N4 O3
Quantity 1 mg
Synonym Paliperidone-(ethyl-D4), 9-Hydroxyrisperidone-(ethyl-D4)
InChI Formula InChI=1S/C23H27FN4O3/c1-14-17(23(30)28-9-2-3-19(29)22(28)25-14)8-12-27-10-6-15(7-11-27)21-18-5-4-16(24)13-20(18)31-26-21/h4-5,13,15,19,29H,2-3,6-12H2,1H3/i8D2,12D2
SMILES [2H]C([2H])(N1CCC(CC1)c2noc3cc(F)ccc23)C([2H])([2H])C4=C(C)N=C5C(O)CCCN5C4=O
IUPAC Name 9-hydroxy-2-methyl-3-[1,1,2,2-tetradeuterio-2-[4-(6-fluoro-1,2-benzoxazol-3-yl)piperidin-1-yl]ethyl]-6,7,8,9-tetrahydropyrido[1,2-a]pyrimidin-4-one
Molecular Weight (g/mol) 430.51
Formula Weight 430.2318
Percent Purity >95