Få mere at vide
Palbociclib N-Aldehyde, TRC
Artikelnummer. 30424581
Skift visning
Mængde:
5 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30424581 10 mg
30418577 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30424581 Leverandør Toronto Research Chemicals Leverandørnr. TRCP13992510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 2174002-16-3
Molekylær formel C25 H29 N7 O3
Molekylvægt (g/mol) 475.54
Synonym 4-(6-((6-Acetyl-8-cyclopentyl-5-methyl-7-oxo-7,8-dihydropyrido[2,3-d]pyrimidin-2-yl)amino)pyridin-3-yl)piperazine-1-carbaldehyde
IUPAC navn 4-[6-[(6-acetyl-8-cyclopentyl-5-methyl-7-oxo-pyrido[2,3-d]pyrimidin-2-yl)amino]-3-pyridyl]piperazine-1-carbaldehyde
SMIL CC(=O)C1=C(C)c2cnc(Nc3ccc(cn3)N4CCN(CC4)C=O)nc2N(C5CCCC5)C1=O
Tekniske data
Kemisk navn eller materiale Palbociclib N-Aldehyde
Mængde 10 mg
InChI formel InChI=1S/C25H29N7O3/c1-16-20-14-27-25(28-21-8-7-19(13-26-21)31-11-9-30(15-33)10-12-31)29-23(20)32(18-5-3-4-6-18)24(35)22(16)17(2)34/h7-8,13-15,18H,3-6,9-12H2,1-2H3,(H,26,27,28,29)
Formel vægt 475.2332