missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human PAK2 (PAK65), GST Tag Recombinant Protein
Human PAK2 (PAK65), GST Tag Recombinant Protein
4375.00 DKK
Specifications
Accession Number | NP_002568.2 |
---|---|
For Use With (Application) | Kinase Assay |
Molecular Weight (g/mol) | 85.5 kDa |
Gene Alias | PAK2 (PAK65), PAK6, PAK delta |
KinaseFamily | STE20 Kinase Family |
Product Code | Brand | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | |||||
10568303
|
Invitrogen
PV4565 |
10μg |
4375.00 DKK
10µg |
Estimated Shipment: 21-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
PAK proteins are critical effectors that link Rho GTPases to cytoskeleton reorganization and nuclear signaling. PAK proteins, a family of serine⁄threonine p21-activating kinases, serve as targets for the small GTP binding proteins Cdc42 and RAC and have been implicated in a wide range of biological activities.Specifications
NP_002568.2 | |
85.5 kDa | |
STE20 Kinase Family | |
Recombinant | |
Storage Buffer: 50 mM Tris pH 7.5, 150 mM NaCl, 0.5 mM EDTA, 0.02% Triton X-100, 2 mM DTT, 50% Glycerol Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
|
Dry Ice | |
Kinases & Inhibitors |
Kinase Assay | |
PAK2 (PAK65), PAK6, PAK delta | |
Human | |
GST Tag | |
Ser⁄Thr Kinases: STE Group | |
Baculovirus |