Få mere at vide
Tocris Bioscience™ PAG 4'-piperazine
Beskrivelse
PAG (phenyl amino glutarimide) 4'-piperazine is a functionalized cereblon ligand for PROTAC™ research and development; incorporates an E3 ligase ligand with a terminal piperazine for onward chemistry to generate Protein Degraders with rigid linkers. Part of a range of functionalized tool molecules for PROTAC R&D.Please contact us for SD files of our available Degrader Building Blocks.
PROTAC™ is a registered trademark of Arvinas Operations, Inc., and is used under license.
Tekniske data
Tekniske data
| Kemisk navn eller materiale | 3-[[4-(1-Piperazinyl)phenyl]amino]-2,6-piperidinedione dihydrochloride |
| CAS | 2922286-32-4 |
| Mængde | 25 mg |
| Mål | Degrader Building Blocks |
| Molekylær formel | C15H20N4O2.2HCl |
| Renhed | 0.95 |