Få mere at vide
Paeoniflorin (>80%), TRC
Artikelnummer. 30425031
Skift visning
Mængde:
50 mg
100 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30425031 100 mg
30427220 250 mg
30421800 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30425031 Leverandør Toronto Research Chemicals Leverandørnr. TRCP133830100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 23180-57-6
Molekylær formel C23 H28 O11
Molekylvægt (g/mol) 480.46
Synonym β-D-Glucopyranoside, (1aR,2S,3aR,5R,5aR,5bS)-5b-[(benzoyloxy)methyl]tetrahydro-5-hydroxy-2-methyl-2,5-methano-1H-3,4-dioxacyclobuta[cd]pentalen-1a(2H)-yl, Peoniflorin (7CI,8CI), β-D-Glucopyranoside, 5b-[(benzoyloxy)methyl]tetrahydro-5-hydroxy-2-methyl-2,5-methano-1H-3,4-dioxacyclobuta[cd]pentalen-1a(2H)-yl, [1aR-(1aα,2β,3aα,5α,5aα,5bα)]-, 2,5-Methano-1H-3,4-dioxacyclobuta[cd]pentalene, β-D-glucopyranoside deriv., (1aR,2S,3aR,5R,5aR,5bS)-5b-[(Benzoyloxy)methyl]tetrahydro-5-hydroxy-2-methyl-2,5-methano-1H-3,4-dioxacyclobuta[cd]pentalen-1a(2H)-yl β-D-glucopyranoside, NSC 178886, Paeoniflorin
SMIL C[C@@]12C[C@@]3(O)O[C@@H](O1)[C@]4(COC(=O)c5ccccc5)[C@H]3C[C@]24O[C@@H]6O[C@H](CO)[C@@H](O)[C@H](O)[C@H]6O
Tekniske data
Kemisk navn eller materiale Paeoniflorin
Renhedsgrad noter HPLC
Mængde 100 mg
InChI formel InChI=1S/C23H28O11/c1-20-9-22(29)13-7-23(20,32-18-16(27)15(26)14(25)12(8-24)31-18)21(13,19(33-20)34-22)10-30-17(28)11-5-3-2-4-6-11/h2-6,12-16,18-19,24-27,29H,7-10H2,1H3/t12-,13-,14-,15+,16-,18+,19-,20+,21+,22-,23+/m1/s1
Formel vægt 480.1632
Procent renhed >95%