Learn More
Kern Sohn Padded Plastic Box for Custom Weight Set
Plastic case for individual weight set.
585.93 DKK - 1476.24 DKK
Specifications
Product Type | Case |
---|---|
Material | Plastic |
For Use With (Equipment) | Class E1 and M3 Weight |
Product Code | Brand | Holds | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Holds | Price | Quantity & Availability | ||||||
17381893
![]() |
Kern Sohn
313-050-400 |
1 mg to 500 g weights |
585.93 DKK
Each |
Estimated Shipment: 05-06-2025
Log in to see stock. |
||||||
17311913
![]() |
Kern Sohn
313-080-400 |
1 mg to 5 kg weights |
1476.24 DKK
Each |
Estimated Shipment: 05-06-2025
Log in to see stock. |
||||||
Description
- Plastic
- For button/compact weight design
Specifications
Case | |
Plastic | |
Class E1 and M3 Weight |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Kern Sohn Padded Plastic Box for Custom Weight Set