missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Paclitaxel EP Impurity N, Mikromol™
Description
Paclitaxel EP Impurity N, Mikromol
Specifications
Specifications
| Kemisk navn eller materiale | Baccatin III |
| CAS | 27548-93-2 |
| Analyt- eller komponentnavne | Baccatin III |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C31 H38 O11 |
| Certificeringer/compliance | ISO 17025 |
| Synonym | 13-O-De[(2R,3S)-3-(benzoylamino)-2-hydroxy-3-phenylpropanoyl]paclitaxel (Baccatin III), Paclitaxel Imp. N (EP), 7,11-Methano-5H-cyclodeca[3,4]benz[1,2-b]oxet-5-one, 6,12b-bis(acetyloxy)-12-(benzoyloxy)-1,2a,3,4,4a,6,9,10,11,12,12a,12b-dodecahydro-4,9,11-trihydroxy-4a,8,13,13-tetramethyl-, (2aR,4S,4aS,6R,9S,11S,12S,12aR,12bS)-, (2aR,4S,4aS,6R,9S,11S,12S,12aR,12bS)-6,12b-Bis(acetyloxy)-12-(benzoyloxy)-1,2a,3,4,4a,6,9,10,11,12,12a,12b-dodecahydro-4,9,11-trihydroxy-4a,8,13,13-tetramethyl-7,11-methano-5H-cyclodeca[3,4]benz[1,2-b]oxet-5-one, Baccatin III, NSC 330753, 7,11-Methano-5H-cyclodeca[3,4]benz[1,2-b]oxet-5-one, 6,12b-bis(acetyloxy)-12-(benzoyloxy)-1,2a,3,4,4a,6,9,10,11,12,12a,12b-dodecahydro-4,9,11-trihydroxy-4a,8,13,13-tetramethyl-, [2aR-(2aalpha,4beta,4abeta,6beta,9alpha,11alpha,12alpha,12aalpha,12balpha)]-, Paclitaxel Impurity N |
| InChI formel | InChI=1S/C31H38O11/c1-15-19(34)13-31(38)26(41-27(37)18-10-8-7-9-11-18)24-29(6,20(35)12-21-30(24,14-39-21)42-17(3)33)25(36)23(40-16(2)32)22(15)28(31,4)5/h7-11,19-21,23-24,26,34-35,38H,12-14H2,1-6H3/t19-,20-,21+,23+,24-,26-,29+,30-,31+/m0/s1 |
| Løsningstype | Neat |
| SMIL | CC(=O)O[C@H]1C(=O)[C@]2(C)[C@@H](O)C[C@H]3OC[C@@]3(OC(=O)C)[C@H]2[C@H](OC(=O)c4ccccc4)[C@]5(O)C[C@H](O)C(=C1C5(C)C)C |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?