Få mere at vide
Paclitaxel-d5 (Benzoyloxy), TRC
Artikelnummer. 30427878
Skift visning
Mængde:
0.5 mg
Artikelnummer. Mængde
30427878 0.5 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30427878 Leverandør Toronto Research Chemicals Leverandørnr. TRCP1325030.5MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Paclitaxel benxoyloxy-d5
CAS 1261254-56-1
Molekylær formel C47 2H5 H46 N O14
Mængde 0.5 mg
Synonym Benzenepropanoic acid','β-(benzoylamino)-α-hydroxy-','(2aR,4S,4aS,6R,9S,11S,12S,12aR,12bS)-6,12b-bis(acetyloxy)-12-(benzoyl-2,3,4,5,6-d5-oxy)-2a,3,4,4a,5,6,9,10,11,12,12a,12b-dodecahydro-4,11-dihydroxy-4a,8,13,13-tetramethyl-5-oxo-7,11-methano-1H-cyclodeca[3,4]benz[1,2-b]oxet-9-yl ester','(αR,βS)-, Paclitaxel-d5, Paclitaxel-D5 (benzoyloxy-D5), (2R,3S)-3-(Benzoylamino)-2-hydroxy-3-phenylpropanoic acid (2aR,4S,4aS,6R,9S,11S,12S,12aR,12bS)-6,12b-bis(acetyloxy)-12-(benzoyl-2,3,4,5,6-d5-oxy)-2a,3,4,4a,5,6,9,10,11,12,12a,12b-dodecahydro-4,11-dihydroxy-4a,8,13,13-tetramethyl-5-oxo-7,11-methano-1H-cyclodeca[3,4]benz[1,2-b]oxet-9-yl ester
InChI formel InChI=1S/C47H51NO14/c1-25-31(60-43(56)36(52)35(28-16-10-7-11-17-28)48-41(54)29-18-12-8-13-19-29)23-47(57)40(61-42(55)30-20-14-9-15-21-30)38-45(6,32(51)22-33-46(38,24-58-33)62-27(3)50)39(53)37(59-26(2)49)34(25)44(47,4)5/h7-21,31-33,35-38,40,51-52,57H,22-24H2,1-6H3,(H,48,54)/t31-,32-,33+,35-,36+,37+,38-,40-,45+,46-,47+/m0/s1/i9D,14D,15D,20D,21D
SMIL [2H]c1c([2H])c([2H])c(C(=O)O[C@H]2[C@@H]3[C@@]4(CO[C@@H]4C[C@H](O)[C@@]3(C)C(=O)[C@H](OC(=O)C)C5=C(C)[C@H](C[C@]2(O)C5(C)C)OC(=O)[C@H](O)[C@@H](NC(=O)c6ccccc6)c7ccccc7)OC(=O)C)c([2H])c1[2H]
Molekylvægt (g/mol) 858.94
Formel vægt 858.3623