missing translation for 'onlineSavingsMsg'
Få mere at vide

Paclitaxel-8-hydro-bicyclo(3.3.0)octane, TRC

Artikelnummer. 30479866
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
5 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30479866 25 mg
30466706 50 mg
30468378 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30479866 Leverandør Toronto Research Chemicals Leverandørnr. TRCM25999025MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 146139-03-9
Molekylær formel C47 H51 N O14
Molekylvægt (g/mol) 853.91
Synonym Benzenepropanoic acid, β-(benzoylamino)-α-hydroxy-, 2a,8-bis(acetyloxy)-3-(benzoyloxy)dodecahydro-4,10-dihydroxy-7,9a,12,12-tetramethyl-9-oxo-3H-4,7a-methanocyclohept[3,3a]indeno[5,4-b]oxet-6-yl ester, [2aS-[2aα,2bS*,3α,4α,6α(αS*,βR*),7β,7aβ,8β,9aβ,10β,11aα]]- (9CI), 3H-4,7a-Methanocyclohept[3,3a]indeno[5,4-b]oxete, benzenepropanoic acid deriv., Paclitaxel-8-hydro-bicyclo(3.3.0)octane, (2R,3S)-3-(Benzoylamino)-2-hydroxy-3-phenylpropanoic acid (1R,2aS,3S,4aR,6aS,6bR,7S,8S,10S,11S,11aR)-1,6a-bis(acetyloxy)-7-(benzoyloxy)dodecahydro-3,8-dihydroxy-2a,11,12,12-tetramethyl-2-oxo-7H-8,11a-methanocyclohept[3,3a]indeno[5,4-b]oxet-10-yl ester, Paclitaxel Photo Degradant, Taxol Photodegradant
SMIL C[C@@H]1[C@H](C[C@@]2(O)[C@@H](OC(=O)c3ccccc3)[C@]45[C@@]6(CO[C@@H]6C[C@H](O)[C@@]4(C)C(=O)[C@H](OC(=O)C)[C@@]15C2(C)C)OC(=O)C)OC(=O)[C@H](O)[C@@H](NC(=O)c7ccccc7)c8ccccc8

Tekniske data

Kemisk navn eller materiale Paclitaxel-8-hydro-bicyclo(3.3.0)octane
Mængde 25 mg
InChI formel InChI=1S/C47H51NO14/c1-25-31(60-40(56)35(52)34(28-16-10-7-11-17-28)48-38(54)29-18-12-8-13-19-29)23-44(57)41(61-39(55)30-20-14-9-15-21-30)47-43(6,32(51)22-33-45(47,24-58-33)62-27(3)50)36(53)37(59-26(2)49)46(25,47)42(44,4)5/h7-21,25,31-35,37,41,51-52,57H,22-24H2,1-6H3,(H,48,54)/t25-,31+,32+,33-,34+,35-,37+,41-,43+,44-,45+,46-,47-/m1/s1
Formel vægt 853.331
Produkttitel
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.