Få mere at vide
Paclitaxel-8-hydro-bicyclo(3.3.0)octane, TRC
Artikelnummer. 30466706
Skift visning
Mængde:
5 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30466706 50 mg
30479866 25 mg
30468378 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30466706 Leverandør Toronto Research Chemicals Leverandørnr. TRCM25999050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 146139-03-9
Molekylær formel C47 H51 N O14
Molekylvægt (g/mol) 853.91
Synonym Benzenepropanoic acid, β-(benzoylamino)-α-hydroxy-, 2a,8-bis(acetyloxy)-3-(benzoyloxy)dodecahydro-4,10-dihydroxy-7,9a,12,12-tetramethyl-9-oxo-3H-4,7a-methanocyclohept[3,3a]indeno[5,4-b]oxet-6-yl ester, [2aS-[2aα,2bS*,3α,4α,6α(αS*,βR*),7β,7aβ,8β,9aβ,10β,11aα]]- (9CI), 3H-4,7a-Methanocyclohept[3,3a]indeno[5,4-b]oxete, benzenepropanoic acid deriv., Paclitaxel-8-hydro-bicyclo(3.3.0)octane, (2R,3S)-3-(Benzoylamino)-2-hydroxy-3-phenylpropanoic acid (1R,2aS,3S,4aR,6aS,6bR,7S,8S,10S,11S,11aR)-1,6a-bis(acetyloxy)-7-(benzoyloxy)dodecahydro-3,8-dihydroxy-2a,11,12,12-tetramethyl-2-oxo-7H-8,11a-methanocyclohept[3,3a]indeno[5,4-b]oxet-10-yl ester, Paclitaxel Photo Degradant, Taxol Photodegradant
SMIL C[C@@H]1[C@H](C[C@@]2(O)[C@@H](OC(=O)c3ccccc3)[C@]45[C@@]6(CO[C@@H]6C[C@H](O)[C@@]4(C)C(=O)[C@H](OC(=O)C)[C@@]15C2(C)C)OC(=O)C)OC(=O)[C@H](O)[C@@H](NC(=O)c7ccccc7)c8ccccc8
Tekniske data
Kemisk navn eller materiale Paclitaxel-8-hydro-bicyclo(3.3.0)octane
Mængde 50 mg
InChI formel InChI=1S/C47H51NO14/c1-25-31(60-40(56)35(52)34(28-16-10-7-11-17-28)48-38(54)29-18-12-8-13-19-29)23-44(57)41(61-39(55)30-20-14-9-15-21-30)47-43(6,32(51)22-33-45(47,24-58-33)62-27(3)50)36(53)37(59-26(2)49)46(25,47)42(44,4)5/h7-21,25,31-35,37,41,51-52,57H,22-24H2,1-6H3,(H,48,54)/t25-,31+,32+,33-,34+,35-,37+,41-,43+,44-,45+,46-,47-/m1/s1
Formel vægt 853.331