Få mere at vide
PAC-1-d8, TRC
Artikelnummer. 30415327
Skift visning
Mængde:
25 mg
Artikelnummer. Mængde
30415327 25 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30415327 Leverandør Toronto Research Chemicals Leverandørnr. TRCP13200225MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale PAC-1-d8
CAS 1287241-26-2
Molekylær formel C23H20D8N4O2
Mængde 25 mg
Synonym 4-(Phenylmethyl)-1-(piperazine-d8)acetic Acid (2E)-2-[[2-Hydroxy-3-(2-propen-1-yl) phenyl]methylene]hydrazide
InChI formel InChI=1S/C23H28N4O2/c1-2-7-20-10-6-11-21(23(20)29)16-24-25-22(28)18-27-14-12-26(13-15-27)17-19-8-4-3-5-9-19/h2-6,8-11,16,29H,1,7,12-15,17-18H2,(H,25,28)/b24-16+
SMIL N#CCCN(C)c1nc2c(cc(OC)c(OC)c2)c(N)n1
Molekylvægt (g/mol) 400.54