missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals p21-Activated Kinase Inhibitor III, IPA-3
Click to view available options
Quantity:
5 mg
10 mg
Description
Acts as an allosteric inhibitor of Pak1
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Quantity | 5 mg |
Description | Yellow |
Content And Storage | -30°C to -10°C |
For Use With (Application) | Acts as an allosteric inhibitor of Pak1 |
Formula Weight | 350.50g/mol |
Sensitivity | Light Sensitive |
Solubility | Soluble in DMSO or ethanol; Insoluble in water |
Product Type | Signal Transduction Reagent |
Form | Solid |
InChI Key | RFAXLXKIAKIUDT-UHFFFAOYSA-N |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'