missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
p19 INK4d Antibody, HRP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-00015H
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
p19 INK4d Polyclonal specifically detects p19 INK4d in Human samples. It is validated for ELISA.
Tekniske data
| p19 INK4d | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| HRP | |
| CDK inhibitor p19INK4d, cell cycle inhibitor, Nur77 associating protein, cyclin-dependent kinase 4 inhibitor D, cyclin-dependent kinase 4 inhibitor D p19, cyclin-dependent kinase inhibitor 2D (p19, inhibits CDK4), inhibitor of cyclin-dependent kinase 4d, INK4D, p19, p19-INK4D | |
| Produced in rabbits immunized with purified, recombinant p19 INK4d (P55273; Met10-Leu166). | |
| 0.1 mL | |
| Cell Cycle and Replication, Ovarian Carcinoma Cell Markers | |
| 1032 | |
| Store at 4°C in the dark. | |
| IgG |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion