Learn More
Brady™ Workstation CLP/GHS Software Download
Align your labels with CLP and GHS classifications using the easy-to-use Workstation CLP/GHS Software.
Brand:  Brady™ 195526
4007.33 DKK valid until 2025-08-24
Use promo code "24132" to get your promotional price.
Product Code. 16303587
  • 4630.00 DKK / Each
Everyday Lab Essentials
Use promo code 24132 at checkout
View Available Promotions
Only null left
Estimated Shipment: 26-06-2025
Description
The Brady™ Workstation CLP/GHS Labels app makes aligning with the CLP Classification, Labeling and Packaging Regulation (1272/2008) and the Globally Harmonized System of Classification and Labeling of Chemicals (GHS) amazingly simple.
  • Easy uploading and storing of Safety Data Sheet (SDS) information and label creation capabilities
  • Download Brady™ Workstation at Workstation.BradyID.com
Specifications
Specifications
Brady Sign and Label Maker, Multicolor and Cut Label Printer, GlobalMark 2 Industrial Label Maker
Windows™ 7, Windows™ 8
Hazard Communication Identification
BWRK-CLP-DWN-EU
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Brady™ Workstation CLP/GHS Software Download > CLP/GHS Software Download; For Use With (Application): Hazard Communication Identification; Operating System: Windows™ 7, Windows™ 8