Få mere at vide
p-Xylene D10, Dr. Ehrenstorfer
Artikelnummer. 30328196
Skift visning
Mængde:
50 mg
Artikelnummer. Mængde
30328196 50 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30328196 Leverandør Dr. Ehrenstorfer Leverandørnr. DRECA17945240
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale p-Xylene-d10
CAS 41051-88-1
Molekylær formel C8 D10
Mængde 50 mg
Synonym 2-Xylene-d10, para-Xylene-d10, 1,4-Dimethylbenzene-d10
InChI formel InChI=1S/C8H10/c1-7-3-5-8(2)6-4-7/h3-6H,1-2H3/i1D3,2D3,3D,4D,5D,6D
SMIL [2H]c1c([2H])c(c([2H])c([2H])c1C([2H])([2H])[2H])C([2H])([2H])[2H]
IUPAC navn 1,2,4,5-tetradeuterio-3,6-bis(trideuteriomethyl)benzene
Molekylvægt (g/mol) 116.23
Formel vægt 116.141 g/mol