Få mere at vide
p-Torasemide (1-(1-Methylethyl)-3-[[4-[(4-methylphenyl)amino]pyridin-3-yl]sulfonyl]urea), Mikromol™
Artikelnummer. 30458478
Skift visning
Mængde:
25 mg
100 mg
4 x 25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30458478 4 x 25 mg
30052209 100 mg
30049465 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30458478 Leverandør Mikromol Leverandørnr. MM1283.0600254AMP
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
p-Torasemide (1-(1-Methylethyl)-3-[[4-[(4-methylphenyl)amino]pyridin-3-yl]sulfonyl]urea), Mikromol
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale p-Torasemide (1-(1-Methylethyl)-3-[(4-[(4-methylphenyl)amino]pyridin-3-yl)sulfonyl]urea)
CAS 106944-63-2
Analyt- eller komponentnavne p-Torasemide (1-(1-Methylethyl)-3-[(4-[(4-methylphenyl)amino]pyridin-3-yl)sulfonyl]urea)
Forsendelsestilstand Room Temperature
Molekylær formel C16 H20 N4 O3 S
Certificeringer/compliance ISO 17025
Synonym p-Torasemide (1-(1-Methylethyl)-3-[[4-[(4-methylphenyl)amino]pyridin-3-yl]sulfonyl]urea), 3-Pyridinesulfonamide, N-[[(1-methylethyl)amino]carbonyl]-4-[(4-methylphenyl)amino]-, 1-Isopropyl-3-((4-m-toluidino-3-pyridyl)sulfonyl)urea, para-Torasemide, p-TRS
InChI formel InChI=1S/C16H20N4O3S/c1-11(2)18-16(21)20-24(22,23)15-10-17-9-8-14(15)19-13-6-4-12(3)5-7-13/h4-11H,1-3H3,(H,17,19)(H2,18,20,21)
Løsningstype Neat
SMIL CC(C)NC(=O)NS(=O)(=O)c1cnccc1Nc2ccc(C)cc2
IUPAC navn 1-[4-(4-methylanilino)pyridin-3-yl]sulfonyl-3-propan-2-ylurea
Molekylvægt (g/mol) 348.42
Mængde 4 x 25 mg
Formel vægt 348.1256 g/mol