Få mere at vide
p-Tolylmercuric Chloride, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | p-Tolylmercuric Chloride |
| CAS | 539-43-5 |
| Molekylær formel | C7 H7 Cl Hg |
| Mængde | 2.5 g |
| Synonym | Mercury','chloro(4-methylphenyl)-, Mercury','chloro-p-tolyl- (7CI,8CI), p-Tolylmercury chloride (6CI), Chloro(4-methylphenyl)mercury, (p-Methylphenyl)mercuric chloride, 4-Methylphenylmercuric chloride, 4-Tolylmercuric chloride, Chloro-p-tolylmercury, Mercuri-p-tolyl chloride, NSC 36721, p-Chloromercuriotoluene, p-Methylphenylmercury chloride, p-Tolylmercuric chloride |
| InChI formel | InChI=1S/C7H7.ClH.Hg/c1-7-5-3-2-4-6-7;;/h3-6H,1H3;1H;/q;;+1/p-1 |
| SMIL | Cc1ccc([Hg]Cl)cc1 |
| IUPAC navn | chloro-(4-methylphenyl)mercury |
| Molekylvægt (g/mol) | 327.17 |
| Formel vægt | 327.9943 |