Få mere at vide
p-Toluidine Trifluoroacetamide, TRC
Artikelnummer. 30452065
Skift visning
Mængde:
500 mg
1 g
2.5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30452065 1 g
30453686 2.5 g
30455974 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30452065 Leverandør Toronto Research Chemicals Leverandørnr. TRCT5362801G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 350-96-9
Molekylær formel C9H8F3NO
Molekylvægt (g/mol) 203.16
Synonym p-Methyltrifluoroacetanilide, 2,2,2-Trifluoro-p-acetotoluidide, 2,2,2-Trifluoro-N-(4-methylphenyl)acetamide, NSC 87411
IUPAC navn 2,2,2-trifluoro-N-(4-methylphenyl)acetamide
SMIL Cc1ccc(NC(=O)C(F)(F)F)cc1
Tekniske data
Kemisk navn eller materiale p-Toluidine Trifluoroacetamide
Mængde 1 g
InChI formel InChI=1S/C9H8F3NO/c1-6-2-4-7(5-3-6)13-8(14)9(10,11)12/h2-5H,1H3,(H,13,14)
Formel vægt 203.0558