Få mere at vide
p-Toluidine Hydrochloride, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | p-Toluidine Hydrochloride |
| CAS | 540-23-8 |
| Molekylær formel | C7 H9 N . H Cl |
| Mængde | 25 g |
| Synonym | Benzenamine','4-methyl-','hydrochloride (1:1) (ACI), Benzenamine','4-methyl-','hydrochloride (9CI), p-Toluidine','hydrochloride (8CI), 4-Aminotoluene hydrochloride, 4-Methylaniline hydrochloride, 4-Methylanilinium chloride, 4-Methylbenzenamine hydrochloride, 4-Methylphenylammonium chloride, p-Methylaniline hydrochloride, p-Methylanilinium chloride, p-Toluidinium chloride |
| InChI formel | InChI=1S/C7H9N.ClH/c1-6-2-4-7(8)5-3-6;/h2-5H,8H2,1H3;1H |
| SMIL | Cl.Cc1ccc(N)cc1 |
| IUPAC navn | 4-methylaniline;hydrochloride |
| Molekylvægt (g/mol) | 143.61 |