Få mere at vide
p-Toluic-d3 Acid (methyl-d3), CDN
Artikelnummer. 30325566
Skift visning
Mængde:
0.1 g
0.25 g
Pakningsstørrelse:
0.1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30325566 0.1 g 0.1g
30313867 0.25 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30325566 Leverandør CDN Leverandørnr. CDND74320.1G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 4-Methylbenzoic Acid D3 (methyl-D3)
CAS 19215-16-8
Molekylær formel C8 D3 H5 O2
Mængde 0.1 g
Synonym Benzoic acid, 4-(methyl-d3)- (9 CI, ACI), 4-(Methyl-d3)benzoic acid (ACI), p-Toluic-α,α,α-d3 acid (8 CI), 4-Methylbenzoic acid D3 (methyl-D3), p-Toluic acid-d3
InChI formel InChI=1 S/C8H8O2/c1-6-2-4-7(5-3-6)8(9)10/h2-5 H,1H3,(H,9,10)/i1D3
SMIL [2 H]C([2 H])([2 H])c1ccc(cc1)C(=O)O
IUPAC navn 4-(trideuteriomethyl)benzoic acid
Molekylvægt (g/mol) 139.166
Formel vægt 139.071 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity